siRNA Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012160.1 | NP_001012160.1 | 2, 3 | 429 | ||
| XM_006253559.2 | XP_006253621.1 | 2, 3 | 284 | |||
| XM_006253561.2 | XP_006253623.1 | 2, 3 | 284 | |||
| GenBank mRNA | BC083717.1 | - | 2, 3 | 429 | 7190699 | |

Gkap1
Genomic Map