siRNA Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012160.1 | NP_001012160.1 | 3, 4 | 578 | ||
| XM_006253559.2 | XP_006253621.1 | 3, 4 | 433 | |||
| GenBank mRNA | BC083717.1 | - | 3, 4 | 578 | 7190699 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gkap1| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012160.1 | NP_001012160.1 | 3, 4 | 578 | ||
| XM_006253559.2 | XP_006253621.1 | 3, 4 | 433 | |||
| GenBank mRNA | BC083717.1 | - | 3, 4 | 578 | 7190699 | |