siRNA Details
| Gene Symbol: Mllt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012162.1 | NP_001012162.1 | 10 | 1548 | ||
| GenBank mRNA | BC087128.1 | - | 10 | 1548 | 7112191 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mllt10| Gene Symbol: Mllt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012162.1 | NP_001012162.1 | 10 | 1548 | ||
| GenBank mRNA | BC087128.1 | - | 10 | 1548 | 7112191 | |