siRNA Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012165.1 | NP_001012165.1 | 5 | 664 | ||
| GenBank mRNA | BC089914.1 | - | 5 | 664 | 7305045 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc8| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012165.1 | NP_001012165.1 | 5 | 664 | ||
| GenBank mRNA | BC089914.1 | - | 5 | 664 | 7305045 | |