siRNA Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012166.1 | NP_001012166.1 | 13 | 2903 | ||
| GenBank mRNA | BC089900.1 | - | 13 | 2903 | 7383307 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb10| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012166.1 | NP_001012166.1 | 13 | 2903 | ||
| GenBank mRNA | BC089900.1 | - | 13 | 2903 | 7383307 | |