siRNA Details
| Gene Symbol: Nap1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012170.1 | NP_001012170.1 | 8, 9 | 723 | ||
| XM_006230823.2 | XP_006230885.1 | 8, 9 | 729 | |||
| XM_006230824.2 | XP_006230886.1 | 9, 10 | 796 | |||
| XM_006230826.2 | XP_006230888.1 | 8, 9 | 729 | |||
| GenBank mRNA | BC085801.1 | - | 8, 9 | 723 | 7110617 | |

Nap1l4
Genomic Map