siRNA Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012176.1 | NP_001012176.1 | 6 | 662 | ||
| GenBank mRNA | BC084704.1 | - | 6 | 662 | 7301243 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsph1| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012176.1 | NP_001012176.1 | 6 | 662 | ||
| GenBank mRNA | BC084704.1 | - | 6 | 662 | 7301243 | |