siRNA Details
| Gene Symbol: LOC100366044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760951.1 | XP_008759173.1 | 1 | 831 | ||
| XM_008760952.1 | XP_008759174.1 | 1 | 831 | |||
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC083807.1 | - | 834 | 7109902 | ||

LOC100366044
Genomic Map