siRNA Details
| Gene Symbol: Sike1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012182.2 | NP_001012182.1 | 4, 5 | 610 | ||
| GenBank mRNA | BC089208.1 | - | 4, 5 | 610 | 7301503 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sike1| Gene Symbol: Sike1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012182.2 | NP_001012182.1 | 4, 5 | 610 | ||
| GenBank mRNA | BC089208.1 | - | 4, 5 | 610 | 7301503 | |