siRNA Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012184.1 | NP_001012184.1 | 5 | 869 | ||
| XM_006234234.2 | XP_006234296.1 | 5 | 626 | |||
| XM_006234235.2 | XP_006234297.1 | 5 | 919 | |||
| XM_006234236.2 | XP_006234298.1 | 5 | 718 | |||
| XM_006234237.2 | XP_006234299.1 | 5 | 1478 | |||
| XM_006234238.2 | XP_006234300.1 | 5 | 626 | |||
| XM_006234239.2 | XP_006234301.1 | 6 | 1116 | |||
| XM_006234241.2 | XP_006234303.1 | 5 | 716 | |||
| XM_008761860.1 | XP_008760082.1 | 5 | 1207 | |||
| GenBank mRNA | BC087094.1 | - | 5 | 869 | 7191334 | |

Rbms1
Genomic Map