siRNA Details
| Gene Symbol: Caprin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012185.2 | NP_001012185.2 | 3 | 318 | ||
| XM_006234652.2 | XP_006234714.1 | 4 | 373 | |||
| GenBank mRNA | AB373991.1 | - | 2 | 233 | ||
| AY155572.1 | - | 2 | 174 | |||
| BC087123.1 | - | 2 | 188 | |||
| CB579393.1 | - | 3 | 318 | |||

Caprin1
Genomic Map