siRNA Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012190.1 | NP_001012190.1 | 7 | 2359 | ||
| XM_008763626.1 | XP_008761848.1 | 6 | 2185 | |||
| GenBank mRNA | BC083594.1 | - | 7 | 2359 | 7132783 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubap1| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012190.1 | NP_001012190.1 | 7 | 2359 | ||
| XM_008763626.1 | XP_008761848.1 | 6 | 2185 | |||
| GenBank mRNA | BC083594.1 | - | 7 | 2359 | 7132783 | |