siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012191.1 | NP_001012191.1 | 5 | 721 | ||
| XM_006239816.2 | XP_006239878.1 | 5 | 660 | |||
| XM_006239817.2 | XP_006239879.1 | 5 | 659 | |||
| XM_006239818.2 | XP_006239880.1 | 5 | 659 | |||
| XM_006239819.2 | XP_006239881.1 | 5 | 660 | |||
| XM_006239822.2 | XP_006239884.1 | 5 | 659 | |||
| GenBank mRNA | BC081889.1 | - | 5 | 721 | 7109493 | |

Dtnb
Genomic Map