siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012191.1 | NP_001012191.1 | 17 | 2722 | ||
| XM_006239816.2 | XP_006239878.1 | 17 | 2661 | |||
| GenBank mRNA | BC081889.1 | - | 17 | 2722 | 7109493 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtnb| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012191.1 | NP_001012191.1 | 17 | 2722 | ||
| XM_006239816.2 | XP_006239878.1 | 17 | 2661 | |||
| GenBank mRNA | BC081889.1 | - | 17 | 2722 | 7109493 | |