siRNA Details
| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012195.1 | NP_001012195.1 | 4 | 465 | ||
| XM_006240772.2 | XP_006240834.1 | 6 | 896 | |||
| XM_006240773.2 | XP_006240835.1 | 6 | 958 | |||
| XM_006240774.2 | XP_006240836.1 | 5 | 804 | |||
| XM_006240775.2 | XP_006240837.1 | 6 | 814 | |||
| XM_006240776.2 | XP_006240838.1 | 5 | 722 | |||
| XM_008765030.1 | XP_008763252.1 | 5 | 1815 | |||
| GenBank mRNA | BC083614.1 | - | 4 | 465 | 7133774 | |

Rnf41
Genomic Map