siRNA Details
| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012200.1 | NP_001012200.1 | 6 | 723 | ||
| XM_006242102.2 | XP_006242164.1 | 6 | 728 | |||
| XM_006242103.2 | XP_006242165.1 | 6 | 774 | |||
| XM_006242104.1 | XP_006242166.1 | 5 | 769 | |||
| XM_008765764.1 | XP_008763986.1 | 5 | 680 | |||
| GenBank mRNA | BC087669.1 | - | 6 | 723 | 7316813 | |

Ttll1
Genomic Map