siRNA Details
| Gene Symbol: Klhdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012203.1 | NP_001012203.1 | 7 | 946 | ||
| XM_006244542.2 | XP_006244604.1 | 6 | 796 | |||
| GenBank mRNA | BC079035.1 | - | 7 | 946 | 7114311 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc3| Gene Symbol: Klhdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012203.1 | NP_001012203.1 | 7 | 946 | ||
| XM_006244542.2 | XP_006244604.1 | 6 | 796 | |||
| GenBank mRNA | BC079035.1 | - | 7 | 946 | 7114311 | |