siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012205.1 | NP_001012205.1 | 18, 19 | 1625 | ||
| XM_008769485.1 | XP_008767707.1 | 12, 13 | 1303 | |||
| GenBank mRNA | AY557199.1 | - | 18, 19 | 1625 | ||
| DQ857324.1 | - | 18, 19 | 1637 | |||
| DQ857325.1 | - | 18, 19 | 1604 | |||

Dpp10
Genomic Map