siRNA Details
| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012206.2 | NP_001012206.1 | 2 | 831 | ||
| GenBank mRNA | BC087038.1 | - | 2 | 817 | 7096388 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phlda3| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012206.2 | NP_001012206.1 | 2 | 831 | ||
| GenBank mRNA | BC087038.1 | - | 2 | 817 | 7096388 | |