siRNA Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012225.2 | NP_001012225.1 | 16 | 1927 | ||
| NM_001160155.1 | NP_001153627.1 | 16 | 1954 | |||
| XM_006244840.2 | XP_006244902.1 | 16 | 1964 | |||
| XM_008767040.1 | XP_008765262.1 | 15 | 1642 | |||
| GenBank mRNA | BC088215.1 | - | 15 | 1885 | 7383944 | |

Mgat4a
Genomic Map