siRNA Details
| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012226.1 | NP_001012226.1 | 9 | 1324 | ||
| GenBank mRNA | BC081938.1 | - | 9 | 1324 | 7115387 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stat4| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012226.1 | NP_001012226.1 | 9 | 1324 | ||
| GenBank mRNA | BC081938.1 | - | 9 | 1324 | 7115387 | |