siRNA Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012267.2 | NP_001012267.1 | 5 | 1073 | ||
| NM_001286969.1 | NP_001273898.1 | 4 | 891 | |||
| XM_011518689.1 | XP_011516991.1 | 5 | 1084 | |||
| GenBank mRNA | AK092603.1 | - | 5 | 888 | ||
| BC071726.1 | - | 5 | 599 | 5554572 | ||
| BX537851.1 | - | 4 | 420 | |||

CENPP
Genomic Map