siRNA Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012267.2 | NP_001012267.1 | 4 | 973 | ||
| NM_001286969.1 | NP_001273898.1 | 3 | 791 | |||
| XM_011518685.2 | XP_011516987.1 | 4 | 984 | |||
| XM_011518689.1 | XP_011516991.1 | 4 | 984 | |||
| XM_017014715.1 | XP_016870204.1 | 4 | 984 | |||
| GenBank mRNA | AK092603.1 | - | 4 | 788 | ||
| AK307435.1 | - | 3 | 311 | |||
| BC071726.1 | - | 4 | 499 | 5554572 | ||
| BX537851.1 | - | 3 | 320 | |||

CENPP
Genomic Map