siRNA Details
| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 2 | 1652 | ||
| GenBank mRNA | AK096490.1 | - | 1 | 97 | ||
| AK309423.1 | - | 2 | 1297 | |||
| BC020441.1 | - | 2 | 1229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOGA3| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 2 | 1652 | ||
| GenBank mRNA | AK096490.1 | - | 1 | 97 | ||
| AK309423.1 | - | 2 | 1297 | |||
| BC020441.1 | - | 2 | 1229 | |||