siRNA Details
| Gene Symbol: Gm3579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY140896.1 | - | 1 | 3691 | ||
| BC066867.1 | - | 1 | 1599 | 30462835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3579| Gene Symbol: Gm3579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY140896.1 | - | 1 | 3691 | ||
| BC066867.1 | - | 1 | 1599 | 30462835 | ||