siRNA Details
| Gene Symbol: Zfp708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012325.2 | NP_001012325.2 | 4 | 2241 | ||
| NM_001012448.2 | NP_001012450.2 | 4 | 2153 | |||
| NM_001012449.2 | NP_001012451.2 | 3 | 2145 | |||
| NM_001292066.1 | NP_001278995.1 | 4 | 2110 | |||
| GenBank mRNA | AK132353.1 | - | 4 | 2115 | ||
| BC089368.1 | - | 4 | 2120 | |||

Zfp708
Genomic Map