siRNA Details
| Gene Symbol: Rxfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012475.1 | NP_001012493.1 | 3 | 232 | ||
| GenBank mRNA | AY906861.1 | - | 3 | 232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxfp2| Gene Symbol: Rxfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012475.1 | NP_001012493.1 | 3 | 232 | ||
| GenBank mRNA | AY906861.1 | - | 3 | 232 | ||