siRNA Details
| Gene Symbol: Wfdc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012704.1 | NP_001012722.1 | 2 | 1017 | ||
| GenBank mRNA | AY542486.1 | - | 2 | 1017 | ||
| AY542487.1 | - | 2 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc13| Gene Symbol: Wfdc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012704.1 | NP_001012722.1 | 2 | 1017 | ||
| GenBank mRNA | AY542486.1 | - | 2 | 1017 | ||
| AY542487.1 | - | 2 | 1015 | |||