siRNA Details
| Gene Symbol: Rcan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012746.1 | NP_001012764.1 | 3 | 500 | ||
| XM_006239218.2 | XP_006239280.1 | 4 | 719 | |||
| GenBank mRNA | AY919305.1 | - | 3 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcan3| Gene Symbol: Rcan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012746.1 | NP_001012764.1 | 3 | 500 | ||
| XM_006239218.2 | XP_006239280.1 | 4 | 719 | |||
| GenBank mRNA | AY919305.1 | - | 3 | 500 | ||