siRNA Details
| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012747.2 | NP_001012765.1 | 6 | 611 | ||
| XM_006242007.2 | XP_006242069.1 | 6 | 887 | |||
| GenBank mRNA | AY917132.1 | - | 6 | 589 | ||
| BC103643.1 | - | 6 | 611 | 7301307 | ||
| FQ227943.1 | - | 6 | 659 | |||

Nol12
Genomic Map