siRNA Details
| Gene Symbol: Cdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012748.1 | NP_001012766.1 | 8 | 1437 | ||
| GenBank mRNA | AB121033.1 | - | 8 | 1437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh20| Gene Symbol: Cdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012748.1 | NP_001012766.1 | 8 | 1437 | ||
| GenBank mRNA | AB121033.1 | - | 8 | 1437 | ||