siRNA Details
| Gene Symbol: NHLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012754.3 | NP_001012772.1 | 7 | 1977 | ||
| NM_001017370.2 | NP_001017370.1 | 6 | 1776 | |||
| NR_073109.1 | 7 | 1719 | ||||
| GenBank mRNA | BC052492.1 | - | 7 | 1657 | ||
| BC140877.1 | - | 7 | 1903 | 9021745 | ||
| BC140878.1 | - | 7 | 1903 | 9021746 | ||
| BX647872.1 | - | 6 | 1666 | |||
| BX648122.1 | - | 7 | 1927 | |||
| CR749318.1 | - | 6 | 1698 | |||

NHLRC3
Genomic Map