siRNA Details
Gene Symbol: LINC00207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028409.1 | 5 | 738 | |||
NR_028410.1 | 4 | 707 | ||||
NR_028411.1 | 4 | 671 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028409.1 | 5 | 738 | |||
NR_028410.1 | 4 | 707 | ||||
NR_028411.1 | 4 | 671 |