siRNA Details
| Gene Symbol: LINC00937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK074886.1 | - | 5 | 1999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00937| Gene Symbol: LINC00937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK074886.1 | - | 5 | 1999 | ||