siRNA Details
| Gene Symbol: C9orf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 2229 | ||
| GenBank mRNA | BC110807.1 | - | 2 | 1620 | 5178248 | |
| BX648492.1 | - | 2 | 1626 | |||
| BX648620.1 | - | 2 | 1641 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf152| Gene Symbol: C9orf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 2229 | ||
| GenBank mRNA | BC110807.1 | - | 2 | 1620 | 5178248 | |
| BX648492.1 | - | 2 | 1626 | |||
| BX648620.1 | - | 2 | 1641 | |||