siRNA Details
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013048.1 | NP_001013066.1 | 4 | 832 | ||
GenBank mRNA | BC086582.1 | - | 4 | 832 | 7307584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013048.1 | NP_001013066.1 | 4 | 832 | ||
GenBank mRNA | BC086582.1 | - | 4 | 832 | 7307584 |