siRNA Details
| Gene Symbol: Aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013061.1 | NP_001013079.1 | 3 | 311 | ||
| GenBank mRNA | FQ228571.1 | - | 3 | 329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aprt| Gene Symbol: Aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013061.1 | NP_001013079.1 | 3 | 311 | ||
| GenBank mRNA | FQ228571.1 | - | 3 | 329 | ||