siRNA Details
| Gene Symbol: Bin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013186.1 | NP_001013204.1 | 8 | 920 | ||
| GenBank mRNA | BC079146.1 | - | 8 | 920 | 7128352 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bin3| Gene Symbol: Bin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013186.1 | NP_001013204.1 | 8 | 920 | ||
| GenBank mRNA | BC079146.1 | - | 8 | 920 | 7128352 | |