siRNA Details
| Gene Symbol: Cbfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013191.1 | NP_001013209.1 | 6 | 1672 | ||
| XM_006255502.2 | XP_006255564.1 | 6 | 1635 | |||
| GenBank mRNA | BC081946.1 | - | 6 | 1672 | 7121452 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbfb| Gene Symbol: Cbfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013191.1 | NP_001013209.1 | 6 | 1672 | ||
| XM_006255502.2 | XP_006255564.1 | 6 | 1635 | |||
| GenBank mRNA | BC081946.1 | - | 6 | 1672 | 7121452 | |