siRNA Details
| Gene Symbol: Clca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013202.2 | NP_001013220.1 | 2 | 281 | ||
| GenBank mRNA | AB119249.1 | - | 2 | 282 | ||
| AB212889.1 | - | 2 | 282 | |||
| AB306511.1 | - | 2 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clca2| Gene Symbol: Clca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013202.2 | NP_001013220.1 | 2 | 281 | ||
| GenBank mRNA | AB119249.1 | - | 2 | 282 | ||
| AB212889.1 | - | 2 | 282 | |||
| AB306511.1 | - | 2 | 282 | |||