siRNA Details
| Gene Symbol: Itgb3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013213.1 | NP_001013231.1 | 5 | 618 | ||
| XM_008763882.1 | XP_008762104.1 | 3 | 435 | |||
| XM_008763883.1 | XP_008762105.1 | 6 | 360 | |||
| XM_008763884.1 | XP_008762106.1 | 5 | 314 | |||
| GenBank mRNA | BC086363.1 | - | 5 | 618 | 7304501 | |

Itgb3bp
Genomic Map