siRNA Details
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013217.1 | NP_001013235.1 | 8, 9 | 1275 | ||
GenBank mRNA | BC083706.1 | - | 8, 9 | 1275 | 7190356 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013217.1 | NP_001013235.1 | 8, 9 | 1275 | ||
GenBank mRNA | BC083706.1 | - | 8, 9 | 1275 | 7190356 |