siRNA Details
| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013222.1 | NP_001013240.1 | 5 | 1278 | ||
| GenBank mRNA | BC089994.1 | - | 5 | 1278 | 7320068 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnd1| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013222.1 | NP_001013240.1 | 5 | 1278 | ||
| GenBank mRNA | BC089994.1 | - | 5 | 1278 | 7320068 | |