siRNA Details
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013228.1 | NP_001013246.1 | 3 | 146 | ||
GenBank mRNA | BC083797.1 | - | 3 | 146 | 7108507 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013228.1 | NP_001013246.1 | 3 | 146 | ||
GenBank mRNA | BC083797.1 | - | 3 | 146 | 7108507 |