siRNA Details
| Gene Symbol: Selplg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013230.1 | NP_001013248.1 | 2 | 462 | ||
| GenBank mRNA | AM778467.1 | - | 2 | 492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selplg| Gene Symbol: Selplg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013230.1 | NP_001013248.1 | 2 | 462 | ||
| GenBank mRNA | AM778467.1 | - | 2 | 492 | ||