siRNA Details
| Gene Symbol: KRBOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129898.1 | NP_001123370.1 | 6 | 598 | ||
| NM_001129899.1 | NP_001123371.1 | 7 | 642 | |||
| NM_001129900.1 | NP_001123372.1 | 7 | 627 | |||
| NM_017776.2 | NP_060246.2 | 6 | 583 | |||
| GenBank mRNA | AK000351.1 | - | 6 | 448 | ||
| AK096413.1 | - | 6 | 490 | |||
| AK097159.1 | - | 1 | 1472 | |||
| AK289703.1 | - | 6 | 480 | |||
| BC012569.1 | - | 6 | 450 | 4283420 | ||
| DA631861.1 | - | 7 | 544 | |||

KRBOX4
Genomic Map