siRNA Details
| Gene Symbol: Gypc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013233.1 | NP_001013251.1 | 3 | 1270 | ||
| XM_006254599.2 | XP_006254661.1 | 3 | 5240 | |||
| GenBank mRNA | AY234182.1 | - | 3 | 1270 | ||
| BC103641.1 | - | 3 | 1183 | 7189007 | ||

Gypc
Genomic Map