siRNA Details
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013238.1 | NP_001013256.1 | 5 | 526 | ||
XM_006256253.2 | XP_006256315.1 | 6 | 716 | |||
GenBank mRNA | BC061813.1 | - | 5 | 526 | 5598663 | |
FQ218172.1 | - | 5 | 538 | |||
FQ228848.1 | - | 5 | 564 |
