siRNA Details
| Gene Symbol: Pttg1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013238.1 | NP_001013256.1 | 6 | 784 | ||
| XM_006256253.2 | XP_006256315.1 | 7 | 974 | |||
| GenBank mRNA | BC061813.1 | - | 6 | 784 | 5598663 | |
| FQ218172.1 | - | 6 | 796 | |||
| FQ228848.1 | - | 6 | 822 | |||

Pttg1ip
Genomic Map