siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013240.2 | NP_001013258.1 | 4 | 504 | ||
| NM_001267054.1 | NP_001253983.1 | 4 | 432 | |||
| NM_001267055.1 | NP_001253984.1 | 3 | 396 | |||
| XM_006238241.2 | XP_006238303.1 | 4 | 520 | |||
| XM_006238242.2 | XP_006238304.1 | 3 | 408 | |||
| XM_008763796.1 | XP_008762018.1 | 4 | 443 | |||
| GenBank mRNA | AY156922.1 | - | 4 | 500 | ||
| BC086343.1 | - | 4 | 493 | 7301739 | ||
| DY317988.1 | - | 3 | 391 | |||
| FM103539.1 | - | 3 | 332 | |||
| FQ217142.1 | - | 3 | 390 | |||

Cdc26
Genomic Map